  • Search Results
  • Single high-dose dexamethasone and sodium salicylate failed to attenuate phosgene-induced acute lung injury in rats.
Single high-dose dexamethasone and sodium salicylate failed to attenuate phosgene-induced acute lung injury in rats.
Toxicology (2013-11-28)
Fangfang Liu, Jürgen Pauluhn, Hubert Trübel, Chen Wang
ABSTRACT
Life-threatening acute lung injury potentially occurs following high-level accidental exposures to phosgene gas. This situation was mirrored in rats exposed nose-only at 900-1000 mg phosgene/m(3)min. At this exposure level, previous studies on rats demonstrated sustained reflexively induced cardiopulmonary dysfunction and evidence of vascular fluid redistribution. These findings challenge the currently applied treatment strategies to mitigate the presumed non-cardiogenic lung edema by steroidal or non-steroidal anti-inflammatory drugs. This study investigates whether high doses of curatively administered dexamethasone (DX; 100 mg/kg bw, ip) and sodium salicylate (SS; 200 mg/kg bw, ip), alone or in combination, show efficacy to mitigate the phosgene-induced lung edema. Exhaled nitric oxide (eNO), animal morbidity and mortality, and increased lung weights one day postexposure served as endpoints of lung injury and drug efficacy. When applying this dosing regimen, SS showed minimal (if any) efficacy while DX, alone or in combination with SS, substantially aggravated the emerging lung edema (lung weights) with 40% mortality. The degree of acute lung injury (ALI) was mirrored by increased eNO. Its direct relationship to ALI-severity was evidenced by decreased eNO following NO-synthetase inhibitor administration (aminoguanidine-aerosol) and associated mitigation of ALI. All non-treated phosgene-exposed as well as treated but non-phosgene-exposed rats survived. This experimental evidence suggests that high-dose corticoid treatments may aggravate the pulmonary toxicity of phosgene. Similarly, this outcome supports the supposition that non-inflammatory, cardiogenic and/or neurogenic factors play a role in this type of acute lung injury.
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone-Water Soluble, suitable for cell culture, BioReagent
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phosgene solution, 15 wt. % in toluene
Sigma-Aldrich
Sodium salicylate, puriss. p.a., ≥99.5% (NT)
Sigma-Aldrich
Sodium salicylate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium salicylate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium salicylate, meets USP testing specifications
Sigma-Aldrich
Sodium salicylate, puriss. p.a., reag. Ph. Eur., 99.5-101.0% (calc. to the dried substance)
Sigma-Aldrich
Phosgene solution, ~20% in toluene
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®