Alanine scanning mutagenesis of hepatitis C virus E2 cysteine residues: Insights into E2 biogenesis and antigenicity.
Virology (2013-12-10)
Wenbo Wang, Mo Guan, Yuan Liu, Qingqiang Xu, Haoran Peng, Xiaoqing Liu, Ziwei Tang, Yongzhe Zhu, Dage Wu, Hao Ren, Ping Zhao, Zhongtian Qi
RESUMEN
Envelope glycoprotein 2 (E2) of hepatitis C virus contains 18 conserved cysteine (Cys) residues in its ectodomain. By cysteine-alanine mutagenesis and function analysis, six Cys in H77 E2 (C494, C508, C552, C564, C607 and C644) were found to be indispensable for recognition by conformation-dependent mAb H53. Removal of any of these Cys residues did not affect E2 heterodimerization with E1, but notably reduced E1E2 transmembrane transportation. These Cys together with C429 and C503 were required for conformation-dependent mAb H48 recognition. All of the above Cys except C607 were required for H77 and Con1 E2 binding to CD81. None of individual mutation of above Cys affected the ability of E2 to induce neutralizing antibodies in mice. Mouse antibodies mainly recognize E2 linear epitopes and are unrelated to epitopes recognized by human E2 antibodies. The findings provide new insights for understanding the biogenesis of functional HCV envelope proteins and HCV neutralizing immunity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
SAFC
L-Cysteine
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Cysteine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Alanine, ≥99.0% (NT)
Supelco
L-Alanine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C