Merck
L-cysteine supplementation as an adjuvant therapy for type-2 diabetes.
Canadian journal of physiology and pharmacology (2012-07-13)
Sushil K Jain
RESUMEN
Diabetes remains a major public health issue. According to the American Diabetes Association, 23.5 million, or 10.7% of people in the USA aged 20 years and older, have diabetes. Type-2 diabetes is treated both by controlling the diet and with oral hypoglycemic drugs. However, for many patients, achieving a tight control of glucose is difficult with current regimens. This chapter discusses a relatively low-cost dietary supplement that could be used as an adjuvant therapy for type-2 diabetes. A review of the literature indicates that cysteine-rich whey protein improves glucose metabolism in diabetic animals and type-2 diabetic patients. Similarly, in animal studies, improvement in glucose metabolism is observed after supplementation with L-cysteine, or molecules containing a cysteine moiety. This chapter discusses the biochemical mechanisms by which L-cysteine can upregulate the insulin-dependent signaling cascades of glucose metabolism. Further studies are needed to examine whether clinical interventions using L-cysteine as an adjuvant therapy indeed help to control glycemia and vascular inflammation in the diabetic patient population.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Supelco
L-Cysteine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
SAFC
L-Cysteine