• Inicio
  • Resultados de la búsqueda
  • Targeted profiling of polar intracellular metabolites using ion-pair-high performance liquid chromatography and -ultra high performance liquid chromatography coupled to tandem mass spectrometry: applications to serum, urine and tissue extracts.
Targeted profiling of polar intracellular metabolites using ion-pair-high performance liquid chromatography and -ultra high performance liquid chromatography coupled to tandem mass spectrometry: applications to serum, urine and tissue extracts.
Journal of chromatography. A (2014-05-28)
Filippos Michopoulos, Nicky Whalley, Georgios Theodoridis, Ian D Wilson, Tom P J Dunkley, Susan E Critchlow
RESUMEN
The effective analysis of polar ionic metabolites by LC-MS, such as those encountered in central carbon metabolism, represents a major problem for metabolic profiling that is not adequately addressed using strategies based on either reversed-phase or HILIC methods. Here we have compared analysis of central carbon metabolites on optimized methods using HILIC, porous graphitic carbon or ion pair chromatography (IPC) using tributyl ammonium as IP reagent. Of the 3 chromatographic approaches examined only IPC enabled us to obtain a robust analytical methodology. This system was used to profile more than a hundred endogenous metabolic intermediates in urine, serum and tissue samples. However, whilst we found IPC to be the best of the approaches examined considerable care was still needed to obtain robust data. Thus, in excess of 40 of representative biological samples were needed to "condition" a new analytical column and further 10 matrix injections were then required at the beginning of each analytical batch in order to obtain robust and reproducible chromatographic separations. An additional limitation that we have found was that, for a small number of phosphorylated and poly carboxylic acid metabolites, measurement was only possible if the analytes were present in relatively high concentrations. We also found that, whilst this methodology could be used for the analysis of both in vitro cell culture media, cell extracts, tissue, and biological fluids (blood, urine), for the best results columns should only be used to analyze a single matrix. However, despite the need for extensive column conditioning, and the manifold disadvantages resulting from the contamination of the separation system and mass spectrometer with the ion pair reagent, IPC-MS currently provides the best means of analyzing these polar, ionic and problematic metabolites.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Supelco
Activated charcoal, powder
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Carbon, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Supelco
Activated charcoal, suitable for GC
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon, foil, 10x10mm, thickness 2.0mm, hOpg
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon, foil, 100x100mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, tube, graphite, 50mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, 99.95%