The aging suppressor klotho: a potential regulator of growth hormone secretion.
American journal of physiology. Endocrinology and metabolism (2014-06-19)
Shiri Shahmoon, Hadara Rubinfeld, Ido Wolf, Zvi R Cohen, Moshe Hadani, Ilan Shimon, Tami Rubinek
RESUMEN
Klotho is a transmembranal protein highly expressed in the kidneys, choroid plexus, and anterior pituitary. Klotho can also be cleaved and shed and acts as a circulating hormone. Klotho-deficient mice (kl/kl mice) develop a phenotype resembling early aging. Several lines of evidence suggest a role for klotho in the regulation of growth hormone (GH) secretion. The kl/kl mice are smaller compared with their wild-type counterparts, and their somatotropes show reduced numbers of secretory granules. Moreover, klotho is a potent inhibitor of the IGF-I pathway, a negative regulator of GH secretion. Therefore, we hypothesized that klotho may enhance GH secretion. The effect of klotho on GH secretion was examined in GH3 rat somatotrophs, cultured rat pituitaries, and cultured human GH-secreting adenomas. In all three models, klotho treatment increased GH secretion. Prolonged treatment of mice with intraperitoneal klotho injections increased mRNA levels of IGF-I and IGF-I-binding protein-3 mRNA in the liver, reflecting increased serum GH levels. In accord with its ability to inhibit the IGF-I pathway, klotho partially restored the inhibitory effect of IGF-I on GH secretion. Klotho is known to be a positive regulator of basic bFGF signaling. We studied rat pituitaries and human adenoma cultures and noted that bFGF increased GH secretion and stimulated ERK1/2 phosphorylation. Both effects were augmented following treatment with klotho. Taken together, our data indicate for the first time that klotho is a positive regulator of GH secretion and suggest the IGF-I and bFGF pathways as potential mediators of this effect.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Somatotropin (STH) from porcine pituitary
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard