• Inicio
  • Resultados de la búsqueda
  • Optimal cultivation of simultaneous ammonium and phosphorus removal aerobic granular sludge in A/O/A sequencing batch reactor and the assessment of functional organisms.
Optimal cultivation of simultaneous ammonium and phosphorus removal aerobic granular sludge in A/O/A sequencing batch reactor and the assessment of functional organisms.
Environmental technology (2014-06-25)
Cuiya Zhang, Hanmin Zhang, Fenglin Yang
RESUMEN
In this study, sequencing batch reactor (SBR) with an anaerobic/aerobic/anoxic operating mode was used to culture granular sludge. Optimal adjustment of cycle duration was achieved by the direction ofpH, oxidation reduction potential and dissolved oxygen parameters. The results showed that the treating efficiency was significantly improved as the cycle was shortened from 450 to 360 min and further to 200 min. Nitrogen and phosphorus removal were nearly quantitative after 50 days operation and maintained stable to the end of the study period. The typical cycle tests revealed that simultaneous denitrification and phosphorus removal occurred when aerobic granules were gradually formed. The nitrite effect tests showed that less than 4.8 mg N/L of the nitrite could enhance superficial specific aerobic phosphate uptake rate (SAPUR) under aerobic condition, indicating that the traditional method to evaluate the capability of total phosphate-accumulating organisms (PAOs) was inaccurate. Additionally, a high level of nitrite was detrimental to PAOs. A novel method was developed to determine the activity of each kind of PAOs and other denitrifying organisms. The results showed that (1) nitrate, besides nitrite, could also enhance SAPUR and (2) aerobic granular sludge could perform denitrification even when phosphate was not supplied under anoxic condition, suggesting that other denitrifying organisms besides denitrifying phosphate-accumulating organisms also contributed to denitrification.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Supelco
Activated charcoal, powder
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Phosphorus, red, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus, red, ≥97.0%
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Carbon, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Supelco
Activated charcoal, suitable for GC
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon, foil, 10x10mm, thickness 2.0mm, hOpg
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon