Sorption behaviors of a persistent toxaphene congener on marine sediments under different physicochemical conditions.
Chemosphere (2014-08-13)
Youssouf Djibril Soubaneh, Jean-Pierre Gagné, Michel Lebeuf, Bruno Gouteux, Vladimir Nikiforov, Mohamed Osman Awaleh
RESUMEN
Sorptive processes are important parameters affecting the mobility, availability and fate of persistent organic pollutants (POPs), such as toxaphene, in aquatic systems. The sorption and desorption behaviors of the B7-1450, a stable toxaphene congener in environment, on marine sediment was studied under different temperature and salinity conditions to better understand the B-1450 distribution in estuarine systems. The data were fitted to different sorption models to characterize sorption behaviors by evaluating sorption coefficients and sequestrated fraction of B7-1450 on sediments. High carbon-normalized sorption coefficients (Koc) of the B7-1450 were observed with values ranging from 3.2×104 to 6.0×104 mL g(-1) under experimental conditions. The data showed an increase of B7-1450 sorption coefficients with the salinity and a decrease with temperature. These investigations indicate that B7-1450 is three times more sequestred on sediments in cold (2°C, 30 psu) than in warm marine conditions (20°C, 30 psu). These results suggest that the mobility and bioavailable of B7-1450 or other POPs from the sediments could be less important in cold marine comparatively in warm marine and warm freshwater media. As a result of climate changes, the warming of mid and high latitudes coastal waters could enhance the mobility of POPs.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Supelco
Activated charcoal, powder
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Carbon, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Supelco
Toxaphene solution, certified reference material, 1000 μg/mL in isooctane
Supelco
Activated charcoal, suitable for GC
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon, foil, 10x10mm, thickness 2.0mm, hOpg
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon, foil, 100x100mm, thickness 0.35mm, flexible graphite, 99.8%