Dissolved organic carbon and relationship with bacterioplankton community composition in 3 lake regions of Lake Taihu, China.
Canadian journal of microbiology (2014-10-11)
Xinghong Pang, Hong Shen, Yuan Niu, Xiaoxue Sun, Jun Chen, Ping Xie
RESUMEN
To clarify the relationships between dissolved organic carbon (DOC) and bacterioplankton community composition (BCC), a 1-year survey (June 2009 - May 2010) was conducted in 3 regions of Lake Taihu (Meiliang Bay, Lake Center, and Eastern Taihu), China. Polymerase chain reaction - denaturing gradient gel electrophoresis was used to analyze the composition and heterogeneity of the bacterioplankton community. Canonical correspondence analysis was used to explore the relationships between DOC concentration and BCC. We found a significant negative correlation between DOC concentration and bacterioplankton community diversity (as measured by the Shannon-Wiener index (H')). The results show that spatial variation in the bacterioplankton population was stronger than the seasonal variation and that DOC concentration influences BCC in Lake Taihu. DOC concentration, followed by macrophyte biomass, water turbidity, and phytoplankton biomass were the most influential factors that account for BCC changes in Lake Taihu. More detailed studies on the relationship between DOC concentration and BCC should focus on differences in DOC concentrations and quality among these lake regions. DOC had a significant impact on BCC in Meiliang Bay. The relationship between DOC and BCC in the 2 other regions studied (Lake Center and Eastern Taihu) was weaker. The results of this study add to our understanding of the BCC in eutrophic lakes, especially regarding the role of the microbial loop in lake ecosystems.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Supelco
Activated charcoal, powder
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Carbon, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Supelco
Activated charcoal, suitable for GC
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon, foil, 10x10mm, thickness 2.0mm, hOpg
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon, foil, 100x100mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, tube, graphite, 50mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, 99.95%