• Inicio
  • Resultados de la búsqueda
  • Feedback of threshold via estimating sources and composition of sedimentary organic matter across trophic gradients in freshwater lakes.
Feedback of threshold via estimating sources and composition of sedimentary organic matter across trophic gradients in freshwater lakes.
The Science of the total environment (2014-09-23)
Xiaoguang Xu, Wei Li, Megumu Fujibayashi, Munehiro Nomura, Takashi Sakamaki, Osamu Nishimura, Xianning Li
RESUMEN
The quantity and quality of sedimentary organic matter (SOM) in relation to material and energy flows are crucial for understanding the current state and future development of lake systems, yet, characterization of organic matter sources and assessment of their relative contributions in different trophic-state lakes caused by anthropogenic impacts are scarcely known. In this study, for obtaining information concerning the source of SOM and its compositional diversity along different trophic gradients, a total of thirty-one sampling sites from four freshwater lakes located in China and Japan were performed by the molecular level analysis using source-specific fatty acid biomarkers. Results indicated that SOM in these lakes was composed of microalgae-, aquatic plant-, terrestrial plant- and bacteria-derived organic matters based on their fatty acid profiles. The scatter plot matrix exhibited correlations between these sources, however, only terrestrial plant-derived organic carbon was a well predictor for sediment TOC with strong, spatiotemporal dynamics. The source and composition of SOM were evidently influenced by lake trophic state with redundancy analysis. Moreover, increase of lake trophic state led to the relatively higher contribution of aquatic organic matter sources to SOM pool compared with terrigenous sources, as evidenced by significant correlations between the trophic state index [TSI (TP)] and the ratio of terrigenous to aquatic fatty acids (TARFA ratio). Yet, this changing trend became more gradual with higher trophic state and prevented the occurrence of regime shift from allochthonous to autochthonous dominant state by a threshold (0.683) of TARFA ratio. Together, a conceptual diagram was proposed, which highlighted the prevailing state of allochthonous source and implicated sedimentary organics in biogeochemistry cycle within freshwater lakes.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Supelco
Activated charcoal, powder
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Carbon, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Supelco
Activated charcoal, suitable for GC
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon, foil, 10x10mm, thickness 2.0mm, hOpg
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon, foil, 100x100mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon