• Inicio
  • Resultados de la búsqueda
  • Metabolite profile analysis reveals association of vitamin B-6 with metabolites related to one-carbon metabolism and tryptophan catabolism but not with biomarkers of inflammation in oral contraceptive users and reveals the effects of oral contraceptives on these processes.
Metabolite profile analysis reveals association of vitamin B-6 with metabolites related to one-carbon metabolism and tryptophan catabolism but not with biomarkers of inflammation in oral contraceptive users and reveals the effects of oral contraceptives on these processes.
The Journal of nutrition (2014-12-21)
Luisa Rios-Avila, Bonnie Coats, Yueh-Yun Chi, Øivind Midttun, Per M Ueland, Peter W Stacpoole, Jesse F Gregory
RESUMEN
The use of oral contraceptives (OCs) has been associated with low plasma pyridoxal 5'-phosphate (PLP). The functional consequences are unclear. To determine whether functional vitamin B-6 insufficiency occurs in OC users and is attributable to OCs, we investigated the associations of PLP with metabolites of one-carbon metabolism, tryptophan catabolism, and inflammation in OC users, and evaluated the effects of OCs on these metabolites. Plasma metabolite concentrations were measured in 157 OC users (20-40 y of age). Associations between PLP and the metabolites were analyzed through use of generalized additive models and partial least squares-discriminant analysis (PLS-DA). Additionally, data from 111 of the 157 OC users were compared to previously reported data from 11 nonusers, at adequate and low vitamin B-6 status, with use of multivariate ANOVA. PLP showed significant (P < 0.05) negative nonlinear association with homocysteine, glutathione, and ratios of asymmetric dimethylarginine to arginine, 3-hydroxykynurenine to 3-hydroxyanthranilic acid, and 3-hydroxykynurenine to kynurenic acid. PLS-DA supported these conclusions and identified 3-hydroxykynurenine and the 3-hydroxykynurenine-to-kynurenine ratio as discriminating biomarkers in women with PLP ≤30 nmol/L. Among the many differences, OC users had significantly higher plasma pyridoxal (157% at adequate and 195% at low vitamin B-6 status), 4-pyridoxic acid (154% at adequate and 300% at low vitamin B-6 status), xanthurenic acid (218% at low vitamin B-6 status), 3-hydroxyanthranilic acid (176% at adequate and 166% at low vitamin B-6 status), quinolinic acid (127% at low vitamin B-6 status), and nicotinamide (197% at low vitamin B-6 status). Biomarkers of inflammation were not associated with PLP, and no differences were found between the 2 groups. PLP is associated with biomarkers of one-carbon metabolism and tryptophan catabolism but not with biomarkers of inflammation in OC users. Independent of vitamin B-6 status, OCs have effects on metabolites and ratios of one-carbon metabolism and tryptophan catabolism but not on biomarkers of inflammation. This study was registered at clinicaltrials.gov as NCT01128244. The study from which data for nonusers was derived was registered as NCT00877812.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Supelco
Activated charcoal, powder
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Kynurenic acid, ≥98%
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Supelco
Tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Supelco
L-Tryptophan, certified reference material, TraceCERT®
Sigma-Aldrich
3-Hydroxy-DL-kynurenine
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Tryptophan, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Cystathionine, ≥90% (HPLC)