• Inicio
  • Resultados de la búsqueda
  • Polybrominated diphenyl ethers in human placenta associated with neonatal physiological development at a typical e-waste recycling area in China.
Polybrominated diphenyl ethers in human placenta associated with neonatal physiological development at a typical e-waste recycling area in China.
Environmental pollution (Barking, Essex : 1987) (2014-12-04)
Long Xu, Xia Huo, Yuling Zhang, Weiqiu Li, Jianqing Zhang, Xijin Xu
RESUMEN
Our aim of this study was to characterize the exposure pattern of polybrominated diphenyl ethers(PBDEs) in human placenta and assess their potential effects on neonates. Placenta samples were obtained from a typical e-waste area in Guiyu and a reference area in Haojiang, China. The median ΣPBDE concentration was 32.25 ng/g lipid weight (lw) in placenta samples from Guiyu, and 5.13 ng/g lw from Haojiang. BDE-209 predominated in placenta samples, followed by BDE-28, -47, -99 -153, -183. Residence in Guiyu contributed the most to elevated PDBE levels. Neonatal physiological indices, including bodymass index (BMI), Apgar 1 score and head circumference, were reduced in Guiyu group. No significant difference was found in neonatal weight between the two groups, but neonatal body length in Guiyu was increased. Our data suggest prenatal exposure to PBDEs is high at the e-waste recycling area, and may lead to adverse physiological development in the fetus.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)