  • Search Results
  • Identification of hydrolytic and isomeric N-oxide degradants of vilazodone by on line LC-ESI-MS/MS and APCI-MS.
Identification of hydrolytic and isomeric N-oxide degradants of vilazodone by on line LC-ESI-MS/MS and APCI-MS.
Journal of pharmaceutical and biomedical analysis (2014-12-03)
Pradipbhai D Kalariya, M V N Kumar Talluri, Prinesh N Patel, R Srinivas
ABSTRACT
The present study reports the degradation behavior of a new antidepressant drug, vilazodone, under various stress conditions as per International Conference on Harmonization guidelines (ICH, Q1A(R2). The investigation involved monitoring decomposition of the drug under hydrolytic (acidic, basic and neutral), oxidative, photolytic and thermal stress conditions and identifying degradation products. A rapid, precise, accurate and robust ultra high performance liquid chromatography (UPLC) method has been developed on a Waters CSH Phenyl-Hexyl column (100 mm × 2.1 mm, 1.7 μm) using gradient elution of 10mM ammonium acetate buffer (pH 5.0) and acetonitrile as mobile phase. The drug was found to be degraded in hydrolytic (acidic and basic) and oxidative conditions, whereas it was stable under neutral hydrolytic, photolytic and thermal stress conditions. The method was extended to quadrupole time-of-flight mass spectrometry (QTOF-MS) for the structural characterization of degradation products. It has been observed that isomeric N-oxide degradation products were formed under oxidative stress condition. The exact location of N-oxidation in the drug was investigated using atmospheric pressure chemical ionization (APCI) due to the formation of characteristic fragment ions. These fragment ions resulted from Meisenheimer rearrangement owing to thermal energy activation at the vaporizer of APCI source. All degradation products were comprehensively characterized by UPLC-ESI-MS/MS and UPLC-APCI-MS experiments. The most probable mechanisms for the formation of degradation products have also been proposed. The method was validated in terms of specificity, linearity, accuracy, precision, and robustness as per ICH guidelines.
Product Description
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ultrapure Acetonitrile
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, suitable for HPLC, contains 10 % (v/v) water, 0.1 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%