Inhibition of AMPK/autophagy potentiates parthenolide-induced apoptosis in human breast cancer cells.
Journal of cellular biochemistry (2014-03-13)
Can Lu, Wenwen Wang, Yongsheng Jia, Xiaodong Liu, Zhongsheng Tong, Binghui Li
RESUMEN
Parthenolide is the main bioactive component in feverfew, a common used herbal medicine, and has been extensively studied in relation to its anti-cancer properties. However there have been very few in-depth studies of the activities of this compound at the molecular level. Here, we showed that parthenolide increased reactive oxygen species (ROS), induced cell death, activated AMPK and autophagy, and led to M phase cell cycle arrest in breast cancer cells. Removal of ROS inhibited all parthenolide-associated events, such as cell death, AMPK activation, autophagy induction, and cell cycle arrest. Blockade of autophagy relieved cell cycle arrest, whereas inhibition of AMPK activity significantly repressed the induction of both autophagy and cell cycle arrest. These observations clearly showed that parthenolide-driven ROS activated AMPK-autophagy pathway. Furthermore, inhibition of either AMPK or autophagy significantly potentiated parthenolide-induced apoptosis. Therefore, our results show that parthenolide activates both apoptosis pathway and AMPK-autophagy survival pathway through the generation of ROS, and that suppression of AMPK or autophagy can potentially enhance the anti-cancer effect of parthenolide on breast cancer cells.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, suitable for cell culture, BioReagent
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Parthenolide, ≥98% (HPLC)
Supelco
Rotenone, PESTANAL®, analytical standard
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard