Downregulated Ku70 and ATM associated to poor prognosis in colorectal cancer among Chinese patients.
OncoTargets and therapy (2014-11-05)
Yuanfang Lu, Jingyan Gao, Yuanming Lu
RESUMEN
Double-strand DNA breaks (DSBs) are a key factor in carcinogenesis. The necessary repair of DSBs is pivotal in maintaining normal cell division. To address the relationship between altered expression of DSB repair of proteins Ku70 and ataxia-telangiectasia mutated (ATM) in colorectal cancer (CRC), we examined the expression levels and patterns of Ku70 and ATM in CRC samples. Expression and coexpression of Ku70 and ATM were investigated by using real-time quantitative polymerase chain reaction assays and confirmed further with fluorescent immunohistochemistry in CRC and pericancerous samples from 112 Chinese patients. Downexpression patterns for both Ku70 and ATM were found in the CRC samples and were significantly associated with advanced tumor node metastasis stage and decreased 5-year overall survival rate. Downregulated Ku70 and ATM were associated with poor disease-free survival. Loss of Ku70 and ATM expression might act as a biomarker to predict poor prognosis in patients with CRC.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
DAPI, for nucleic acid staining
Supelco
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone GG-7, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone HP-6017, ascites fluid
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal, clone HP-6017, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Goat/Sheep IgG−Peroxidase antibody, Mouse monoclonal antibody produced in mouse, clone GT-34, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)–Peroxidase antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
Sigma-Aldrich
Anti-Human IgG (Fc specific)−FITC antibody, Mouse monoclonal antibody produced in mouse, clone HP-6017, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Goat/Sheep IgG antibody produced in mouse, clone GT-34, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG, Native−Peroxidase antibody, Mouse monoclonal antibody produced in mouse, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Human IgG (Fab specific) antibody produced in mouse, clone GG-6, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG, Native antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Human IgG2 (Fc specific) antibody produced in mouse, clone HP-6002, ascites fluid
Sigma-Aldrich
Anti-Goat/Sheep IgG−Biotin antibody, Mouse monoclonal, clone GT-34, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Goat/Sheep IgG−Alkaline Phosphatase antibody, Mouse monoclonal, clone GT-34, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Goat/Sheep IgG−FITC antibody, Mouse monoclonal antibody produced in mouse, clone GT-34, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Human IgG4 (pFc′ specific) antibody produced in mouse, clone HP-6023, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Human IgG (γ-chain specific) antibody produced in mouse, clone GG-5, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Rat IgG2a antibody produced in mouse, clone R2A-2, ascites fluid