  • Search Results
  • A new spiroplasma isolate from the field cricket (Gryllus bimaculatus) in Taiwan.
A new spiroplasma isolate from the field cricket (Gryllus bimaculatus) in Taiwan.
Journal of invertebrate pathology (2014-05-08)
Yu-Shin Nai, Ping-Yi Su, Yu-Hsiang Hsu, Ching-Hao Chiang, Jae Su Kim, Yue-Wen Chen, Chung-Hsiung Wang
ABSTRACT
We briefly described the morphology and transmission pathway of a Spiroplasma sp. isolated from the field cricket, Gryllus bimaculatus in Taiwan, followed by the phylogenetic analysis based on the 16S rRNA gene sequence. The cricket spiroplasma infected the hemolymph, gut, muscle tissues and tracheal cells; therefore we suggest that the pathogen invaded tissues and organs from the hemolymph through the tracheal system and the endoplasmic reticular system. Based on 16S rRNA gene sequences and the phylogeny, this spiroplasma was most closely related to Spiroplasma platyhelix (Identity=95%) isolated from the dragonfly Pachydiplax longipennis and belongs to the Ixodetis clade.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, suitable for HPLC, contains 10 % (v/v) water, 0.1 % (v/v) trifluoroacetic acid
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%