Rapid control of severe neoplastic hypercortisolism with metyrapone and ketoconazole.
European journal of endocrinology (2015-01-28)
Jean-Benoît Corcuff, Jacques Young, Pauline Masquefa-Giraud, Philippe Chanson, Eric Baudin, Antoine Tabarin
RESUMEN
Severe Cushing's syndrome elicited by ectopic ACTH syndrome (EAS) or adrenal carcinoma (ACC) can threaten life in the short term. The effectiveness of oral administration of the inhibitors of steroidogenesis ketoconazole and metyrapone in this situation is poorly described. To report the short-term effectiveness and tolerability of metyrapone and ketoconazole elicited either by EAS or by ACC in patients exhibiting severe hypercortisolism. Retrospective analysis of data obtained for patients with urinary free cortisol (UFC) level estimated to be fivefold the upper limit of the normal range (ULN). A total of 14 patients with EAS and eight with ACC treated in two tertiary-care university hospitals. Metyrapone and ketoconazole treatment in combination (along with symptomatic treatments for co-morbidities). Evolution of clinically relevant endpoints (blood pressure, kalaemia and glycaemia) and biological intensity of hypercortisolism 1 week and 1 month after starting steroidogenesis inhibition. After 1 week of treatment, median UFC fell from 40.0 to 3.2 ULN and from 16.0 to 1.0 ULN in patients with EAS and ACC respectively. Median UFC after 1 month of treatment was 0.5 and 1.0 ULN in patients with EAS and ACC respectively and UFC values were normal in 73 and 86% of patients respectively. Clinical status improved dramatically along with kalaemia, glycaemia and blood pressure, allowing a decrease in the relevant treatments.Side effects were minimal and only two patients (one EAS and one ACC) experienced plasma transaminase elevations necessitating ketoconazole withdrawal. Metyrapone-ketoconazole combination therapy is well tolerated and provides rapid control of endocrine cancer-related life-threatening hypercortisolism.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Supelco
2,4′-DDD, PESTANAL®, analytical standard
Sigma-Aldrich
Ketoconazole
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Metyrapone, ≥98% (HPLC), solid
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Methyl-1,2-di-3-pyridyl-1-propanone, 96%
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard