Effects of carbon-ion beam irradiation on the angiogenic response in lung adenocarcinoma A549 cells.
Cell biology international (2014-06-20)
Yuanyuan Liu, Yang Liu, Hong Zhang, Chao Sun, Qiuyue Zhao, Cixia Di, Hongyan Li, Lu Gan, Yali Wang
RESUMEN
Radiotherapy has been focused mainly on killing cancer cells, and little attention has been paid to the process supporting tumor growth and metastasis, including the process of angiogenesis. To investigate the effects of carbon-ion irradiation on angiogenesis in lung cancer cells, we examined the expression of vascular endothelial growth factor and basic fibroblast growth factor in the tumor conditioned medium (TCM) of A549 cells exposed to carbon-ion or X-ray irradiation, as well as endothelial cell growth, invasion, and tube formation induced by TCM. No changes in vascular endothelial growth factor secretion were detected in the TCM of A549 cells exposed to carbon-ion irradiation at 2 or 4 Gy, whereas 1 Gy of irradiation significantly decreased vascular endothelial growth factor and basic fibroblast growth factor levels. Carbon-ion irradiation at 1 Gy inhibited endothelial cell invasion and tube formation. The TCM from A549 cells irradiated with X-ray promoted angiogenesis, whereas the TCM of A549 cells exposed to carbon-ion irradiation at 2 or 4 Gy had no effect. These findings suggest that carbon-ion irradiation at 1 Gy significantly suppressed the process of angiogenesis in vitro by inhibiting endothelial cell invasion and tube formation, which are related to vascular endothelial growth factor and basic fibroblast growth factor production.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard