  • Search Results
  • Zinc deficiency in rats is associated with up-regulation of hippocampal NMDA receptor.
Zinc deficiency in rats is associated with up-regulation of hippocampal NMDA receptor.
Progress in neuro-psychopharmacology & biological psychiatry (2014-10-08)
Urszula Doboszewska, Magdalena Sowa-Kućma, Katarzyna Młyniec, Bartłomiej Pochwat, Malgorzata Hołuj, Beata Ostachowicz, Andrzej Pilc, Gabriel Nowak, Bernadeta Szewczyk
ABSTRACT
Data indicated that zinc deficiency may contribute to the development of depression; however changes induced by zinc deficiency are not fully described. In the present paper we tested whether the dietary zinc restriction in rats causes alterations in N-methyl-D-aspartate receptor (NMDAR) subunits in brain regions that are relevant to depression. Male Sprague Dawley rats were fed a zinc adequate diet (ZnA, 50 mg Zn/kg) or a zinc deficient diet (ZnD, 3 mg Zn/kg) for 4 or 6weeks. Then, the behavior of the rats was examined in the forced swim test, sucrose intake test and social interaction test. Western blot assays were used to study the alterations in NMDAR subunits GluN2A and GluN2B and proteins associated with NMDAR signaling in the hippocampus (Hp) and prefrontal cortex (PFC). Following 4 or 6 weeks of zinc restriction, behavioral despair, anhedonia and a reduction of social behavior occurred in rats with concomitant increased expression of GluN2A and GluN2B and decreased expression of the PSD-95, p-CREB and BDNF protein levels in the Hp. The up-regulation of GluN2A protein was also found in the PFC, but only after prolonged (6 weeks) zinc deprivation. The procedure of zinc restriction in rats causes behavioral changes that share some similarities to the pathophysiology of depression. Obtained data indicated that depressive-like behavior induced by zinc deficiency is associated with the changes in NMDAR signaling pathway.
Product Description
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, ACS reagent, 70%