  • Search Results
  • Characterization of different carbon nanotubes for the development of a mucoadhesive drug delivery system for intravesical treatment of bladder cancer.
Characterization of different carbon nanotubes for the development of a mucoadhesive drug delivery system for intravesical treatment of bladder cancer.
International journal of pharmaceutics (2015-01-18)
Christiane Rieger, David Kunhardt, Anika Kaufmann, Darja Schendel, Doreen Huebner, Kati Erdmann, Stefan Propping, Manfred P Wirth, Bernd Schwenzer, Susanne Fuessel, Silke Hampel
ABSTRACT
In order to increase the effectiveness of therapeutics for bladder carcinoma (BCa) treatment, alternative strategies for intravesical applications are needed. The use of carbon nanotubes (CNTs) as basis for a multifunctional drug transporter is a promising possibility to combine traditional chemotherapeutics with innovative therapeutic agents such as antisense oligodeoxynucleotides or small interfering RNA. In the current study four CNT types varying in length and diameter (CNT-1, CNT-2, CNT-3, CNT-4) were synthesized and then characterized with different spectroscopic techniques. Compared to the pristine CNT-1 and CNT-3, the shortened CNT-2 and CNT-4 exhibited more defects and lower aspect ratios. To analyze their mucoadhesive properties, CNTs were exposed to mouse bladders ex vivo by using Franz diffusion cells. All four tested CNT types were able to adhere to the urothelium with a mean covering area of 5-10%. In vitro studies on UM-UC-3 and EJ28 BCa cells were conducted to evaluate the toxic potential of these CNTs. Viability and cytotoxicity assays revealed that the shortened CNT-2 and CNT-4 induced stronger inhibitory effects on BCa cells than CNT-1 and CNT-3. In conclusion, CNT-1 and CNT-3 showed the most promising properties for further optimization of a multifunctional drug transporter.
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications