  • Search Results
  • Simultaneous detection of 93 conventional and emerging drugs of abuse and their metabolites in urine by UHPLC-MS/MS.
Simultaneous detection of 93 conventional and emerging drugs of abuse and their metabolites in urine by UHPLC-MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-09-10)
Magdalene H Y Tang, C K Ching, Caroline Y W Lee, Ying-Hoo Lam, Tony W L Mak
ABSTRACT
Novel psychoactive substances (NPS) are becoming increasingly popular worldwide in recent years, some of which have been reported to cause considerable harm and even fatalities. Currently, simultaneous screening for a comprehensive panel of conventional and novel drugs of abuse is not widely available in most clinical laboratories. The aim of this study was to establish a chromatography/mass spectrometry-based analytical system for the simultaneous detection of conventional drugs of abuse and NPS in urine. Sample preparation entails enzyme digestion and solid phase extraction; analytes were then detected by liquid-chromatography tandem mass spectrometry (LC-MS/MS) with multiple reaction monitoring. Forty-seven conventional drugs (28 parent drugs, 19 metabolites) and 46 NPS analytes (44 parent drugs, two metabolites) are covered by the established method, which has been validated according to international guidelines. The method was then applied to 964 urine samples collected from drug abusers and the results revealed the presence of two NPS - TFMPP and methcathinone - as well as conventional drugs of abuse. To conclude, an LC-MS/MS method has been established that allows the simultaneous detection of over 90 conventional as well as novel psychoactive substances and metabolites in urine samples. The method was successfully applied to authentic specimens revealing the presence of conventional as well as novel drugs of abuse in the local population.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Sodium acetate solution, NMR reference standard, 50 mM in D2O (99.9 atom % D), water 1 %, NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Millipore
Bifido Selective Supplement B, suitable for microbiology