• Inicio
  • Resultados de la búsqueda
  • Bioassay-guided preparative separation of angiotensin-converting enzyme inhibitory C-flavone glycosides from Desmodium styracifolium by recycling complexation high-speed counter-current chromatography.
Bioassay-guided preparative separation of angiotensin-converting enzyme inhibitory C-flavone glycosides from Desmodium styracifolium by recycling complexation high-speed counter-current chromatography.
Journal of pharmaceutical and biomedical analysis (2014-12-03)
Ying-Qi Zhang, Jian-Guang Luo, Chao Han, Jin-Fang Xu, Ling-Yi Kong
RESUMEN
A new strategy of the convergence of high-speed counter-current chromatography (HSCCC) and bioactive assay technique was developed for rapidly screening and separating the angiotensin-converting enzyme (ACE) inhibitors from the aerial parts of Desmodium styracifolium. Bioactivity-guided fractionation of the crude extract was first established to target the bioactive fractions based on HSCCC coupled with in vitro ACE inhibitory assay. Subsequently, the bioactive fractions were further separated by the recycling complexation HSCCC respectively, using 0.10 mol/L copper sulfate in the lower phase of two-phase solvent system composed of n-butanol/water (1:1, v/v). Five C-glycosylflavones, vicenin 2 (1), carlinoside (2), vicenin 1 (3), schaftoside (4) and vicenin 3 (5), were successfully obtained. Their chemical structures were identified using ESI-MS and NMR. All the isolates showed in vitro ACE inhibitory activity with the IC50 values between 33.62 and 58.37 μM. The results demonstrated that the established method was proposed as an excellent strategy to systematically screen and purify active compounds from traditional Chinese medicines.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder (may contain lumps)
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Phthaldialdehyde, for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Octylphosphonic acid, 97%
Supelco
Acetonitrile, analytical standard