  • Search Results
  • Berberine derivatives reduce atherosclerotic plaque size and vulnerability in apoE(-/-) mice.
Berberine derivatives reduce atherosclerotic plaque size and vulnerability in apoE(-/-) mice.
Journal of translational medicine (2014-11-27)
Junwen Chen, Jiatian Cao, Lu Fang, Bo Liu, Qing Zhou, Yinggang Sun, Yue Wang, Yigang Li, Shu Meng
ABSTRACT
Our previous in vitro and clinical work has demonstrated anti-inflammatory effects of berberine (BBR), but the clinical application of BBR is limited by its poor bioavailability. Derivatives of BBR have been suggested to have enhanced bioavailability compared to BBR. In this study, we tested whether BBR derivatives, compared with BBR, had superior beneficial effects on atherosclerotic plaques in apoE(-/-) mice, and defined possible molecular mechanisms underlying such effects. Macrophages were pretreated with BBR and its derivatives, dihydroberberine (dhBBR) and 8,8-dimethyldihydroberberine (Di-MeBBR), before incubation with oxLDL. Cell surface EMMPRIN expression was measured by flow cytometry and Western blotting, and phospho-(p)-p38, p-JNK, nuclear NFκB p65, and phospho-p65 were measured by Western blotting. ApoE-/- mice fed with the Western diet for 16 weeks were treated with BBR, dhBBR and Di-MeBBR 16 weeks. Aortic atherosclerotic lesion size, plaque matrix proteins, and EMMPRIN and other inflammatory factors were measured using Oil Red O Staining, Masson's trichromestaining and immunohistochemical staining and real-time PCR. Compared with BBR, dhBBR and Di-MeBBR significantly reduced EMMPRIN expression, which was associated with a greater inhibition of p-p38, p-JNK, nuclear NFκB p65 and phospho-p65 induced by oxLDL in macrophages. dhBBR and Di-MeBBR, but not BBR, reduced atherosclerotic plaque size and improved plaque stability indicated by increased α-smooth muscle actin and collagen content, and thicker fibrous caps. dhBBR and Di-MeBBR reduced expression of EMMPRIN, CD68, and NFκB p65, and Di-MeBBR also reduced expression of matrix metalloproteinase-9, intercellular adhesion molecule-1, and vascular cell adhesion molecule-1 in aortic plaques. These results have demonstrated that BBR derivatives, dhBBR and Di-MeBBR, are superior to BBR in inhibiting inflammation and reducing plaque size and vulnerability.
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, suitable for plant cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications