Child care and cortisol across early childhood: context matters.
Developmental psychology (2013-06-19)
Daniel Berry, Clancy Blair, Alexandra Ursache, Michael Willoughby, Patricia Garrett-Peters, Lynne Vernon-Feagans, Mary Bratsch-Hines, W Roger Mills-Koonce, Douglas A Granger
RESUMEN
A considerable body of literature suggests that children's child-care experiences may impact adrenocortical functioning in early childhood. Yet emerging findings also suggest that the magnitude and sometimes the direction of child-care effects on development may be markedly different for children from higher risk contexts. Using data from a large population-based sample of families from predominantly low-income backgrounds in rural communities, we tested the degree to which links between children's child-care experiences (at 7-36 months) and their subsequent cortisol levels (at 48 months) were moderated by their level of cumulative environmental risk. Our results provided evidence of a crossover interaction between cumulative risk and child-care quantity. For children from low-risk contexts, greater weekly hours in child care were predictive of higher cortisol levels. In contrast, for children facing several cumulative risk factors, greater hours in child care per week were predictive of lower cortisol levels. These effects were robust after adjusting for several controls, including children's cortisol levels in early infancy. Child-care quality and type were not predictive of children's cortisol levels, and neither mitigated the conditional effect of child-care quantity on cortisol. These findings suggest that links between child care and children's development may differ as a function of children's broader ecologies.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard