Ginsenoside Rc modulates Akt/FoxO1 pathways and suppresses oxidative stress.
Archives of pharmacal research (2013-08-07)
Dae Hyun Kim, Chan Hum Park, Daeui Park, Yeon Ja Choi, Min Hi Park, Ki Wung Chung, So Ra Kim, Jun Sik Lee, Hae Young Chung
RESUMEN
Ginsenoside Rc (Rc), a protopanaxadiol type ginsenoside, is the active component mainly responsible for the therapeutic and pharmacologic properties of ginseng, which are derived from its suppression of superoxide-induced free radicals. Forkhead box O (FoxO1) regulates various genes involved in cellular metabolism related to cell death and response to oxidative stress, and Rc is known to prevent FoxO1 phosphorylation by activation of PI3K/Akt and subsequent inhibition of AMP-activated protein kinase (AMPK) in cells exposed to tert-butylhydroperoxide (t-BHP). In the current study, we attempted the mechanism of increased catalase expression by Rc through inhibition of FoxO1 activation resulting from t-BHP-induced production of reactive species (RS). We found that overexpression of catalase induced by Rc resulted in suppression of RS production in kidney human embryo kidney 293T cells (HEK293T) cells, and that oxidative stress induced activation of PI3K/Akt and inhibition of the AMPK pathway and FoxO1 phosphorylation, leading to down-regulation of catalase, a FoxO1-targeting gene. In addition, treatment of HEK293T cells with Rc resulted in cAMP-response element-binding protein (CREB)-binding protein (CBP) regulated FoxO1 acetylation. Our results suggest that Rc modulates FoxO1 phosphorylation through activation of PI3K/Akt and inhibition of AMPK and FoxO1 acetylation through interaction with CBP and SIRT1, and that this leads to upregulation of catalase under conditions of oxidative stress.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Fluorescein diacetate, used as cell viability stain
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)–Peroxidase antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG, Native−Peroxidase antibody, Mouse monoclonal antibody produced in mouse, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG, Native antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Diacetylfluorescein, European Pharmacopoeia (EP) Reference Standard