• Inicio
  • Resultados de la búsqueda
  • The transcription factor GTF2IRD1 regulates the topology and function of photoreceptors by modulating photoreceptor gene expression across the retina.
The transcription factor GTF2IRD1 regulates the topology and function of photoreceptors by modulating photoreceptor gene expression across the retina.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-11-14)
Tomohiro Masuda, Xiaodong Zhang, Cindy Berlinicke, Jun Wan, Anitha Yerrabelli, Elizabeth A Conner, Sten Kjellstrom, Ronald Bush, Snorri S Thorgeirsson, Anand Swaroop, Shiming Chen, Donald J Zack
RESUMEN
The mechanisms that specify photoreceptor cell-fate determination, especially as regards to short-wave-sensitive (S) versus medium-wave-sensitive (M) cone identity, and maintain their nature and function, are not fully understood. Here we report the importance of general transcription factor II-I repeat domain-containing protein 1 (GTF2IRD1) in maintaining M cone cell identity and function as well as rod function. In the mouse, GTF2IRD1 is expressed in cell-fate determined photoreceptors at postnatal day 10. GTF2IRD1 binds to enhancer and promoter regions in the mouse rhodopsin, M- and S-opsin genes, but regulates their expression differentially. Through interaction with the transcription factors CRX and thyroid hormone receptor β 2, it enhances M-opsin expression, whereas it suppresses S-opsin expression; and with CRX and NRL, it enhances rhodopsin expression. In an apparent paradox, although GTF2IRD1 is widely expressed in multiple cell types across the retina, knock-out of GTF2IRD1 alters the retinal expression of only a limited number of annotated genes. Interestingly, however, the null mutation leads to altered topology of cone opsin expression in the retina, with aberrant S-opsin overexpression and M-opsin underexpression in M cones. Gtf2ird1-null mice also demonstrate abnormal M cone and rod electrophysiological responses. These findings suggest an important role for GTF2IRD1 in regulating the level and topology of rod and cone gene expression, and in maintaining normal retinal function.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)–Peroxidase antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG, Native−Peroxidase antibody, Mouse monoclonal antibody produced in mouse, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG, Native antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture