  • Search Results
  • Systemic exposure to and disposition of catechols derived from Salvia miltiorrhiza roots (Danshen) after intravenous dosing DanHong injection in human subjects, rats, and dogs.
Systemic exposure to and disposition of catechols derived from Salvia miltiorrhiza roots (Danshen) after intravenous dosing DanHong injection in human subjects, rats, and dogs.
Drug metabolism and disposition: the biological fate of chemicals (2015-02-12)
Meijuan Li, Fengqing Wang, Yühong Huang, Feifei Du, Chenchun Zhong, Olajide E Olaleye, Weiwei Jia, Yanfen Li, Fang Xu, Jiajia Dong, Jian Li, Justin B R Lim, Buchang Zhao, Lifu Jia, Li Li, Chuan Li
ABSTRACT
DanHong injection is a Danshen (Salvia miltiorrhiza roots)-based injectable solution for treatment of coronary artery disease and ischemic stroke. Danshen catechols are believed to be responsible for the injection's therapeutic effects. This study aimed to characterize systemic exposure to and elimination of Danshen catechols in human subjects, rats, and dogs receiving intravenous DanHong injection. A total of 28 catechols were detected, with content levels of 0.002-7.066 mM in the injection, and the major compounds included tanshinol, protocatechuic aldehyde, salvianolic acid B, rosmarinic acid, salvianolic acids A and D, and lithospermic acid with their daily doses ≥10 μmol/subject. After dosing, tanshinol, salvianolic acid D, and lithospermic acid exhibited considerable exposure in human subjects and rats. However, only tanshinol had considerable exposure in dogs. The considerable exposure to tanshinol was due to its having the highest dose, whereas that to salvianolic acid D and lithospermic acid was due to their relatively long elimination half-lives in the human subjects and rats. Protocatechuic aldehyde and rosmarinic acid circulated in the bloodstream predominantly as metabolites; salvianolic acids A and B exhibited low plasma levels with their human plasma metabolites little or not detected. Tanshinol and salvianolic acid D were eliminated mainly via renal excretion. Elimination of other catechols involved hepatobiliary and/or renal excretion of their metabolites. Methylation was found to be the primary metabolism for most Danshen catechols and showed intercompound and interspecies differences in rate and degree in vitro. The information gained here is relevant to pharmacological and toxicological research on DanHong injection.
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Isovanillin, ≥95.0%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Supelco
3,4-Dihydroxybenzoic acid, analytical standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
SAFC
Tromethamine
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Protocatechuic acid, primary reference standard
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
Trometamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Supelco
Chrysin, analytical standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, suitable for HPLC, contains 10 % (v/v) water, 0.1 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Chrysin, 97%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline