  • Search Results
  • Folate-grafted boron nitride nanotubes: possible exploitation in cancer therapy.
Folate-grafted boron nitride nanotubes: possible exploitation in cancer therapy.
International journal of pharmaceutics (2015-02-01)
Tiago H Ferreira, Attilio Marino, Antonella Rocca, Ioannis Liakos, Simone Nitti, Athanassia Athanassiou, Virgilio Mattoli, Barbara Mazzolai, Edesia M B de Sousa, Gianni Ciofani
ABSTRACT
Boron nitride nanotubes (BNNTs) have generated considerable interest among the scientific community because of their unique physical and chemical properties. They present good chemical inertness, high thermal stability, and optimal resistance to oxidation, that make them ideal candidates for biomedical applications, in particular as nanovectors for drug, gene and protein delivery into the cells. In this study, BNNTs were prepared through a synthesis based on a chemical vapor deposition (CVD) method, and thereafter chemically functionalized with folic acid. The obtained nanostructures have been characterized by Fourier transform infrared spectroscopy (FTIR), X-ray photoelectron spectroscopy (XPS), thermogravimetric analysis (TGA), scanning electron microscopy (SEM), and transmission electron microscopy (TEM). The characterization showed efficiently functionalized BNNTs of length of about 1 μm. Furthermore, confocal laser microscopy demonstrated that our nanotubes can be fluorescently-traced under appropriate excitation. Thanks to this property, it has been possible to investigate their internalization by HeLa cells through confocal microscopy, demonstrating that the BNNT up-take clearly increases after the functionalization with folate, a result confirmed by inductively coupled plasma (ICP) assessment of boron content inside the treated cell cultures.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Hydrogen chloride, puriss., ≥99.8%
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Triton X-100, laboratory grade
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Boron, crystalline, 1 cm, 99.7% trace metals basis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Boron, ≥95% (boron), amorphous powder
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Boron, microfoil, disks, 10mm, thinness 0.1μm, specific density 23.5μg/cm2, permanent mylar 3.5μm support, 99.6%
Boron, monofilament, 50m, diameter 0.1mm
Boron, monofilament, 10m, diameter 0.2mm
Boron, microfoil, disks, 25mm, thinness 0.1μm, specific density 23.5μg/cm2, permanent mylar 3.5μm support, 99.6%
Boron, monofilament, 10m, diameter 0.1mm
Boron, monofilament, 100m, diameter 0.1mm
Boron, monofilament, 5m, diameter 0.1mm
Boron, monofilament, 20m, diameter 0.1mm
Boron, monofilament, 200m, diameter 0.1mm
Boron, monofilament, 5m, diameter 0.2mm