Influence of trypsinization and alternative procedures for cell preparation before RNA extraction on RNA integrity.
Analytical biochemistry (2014-07-02)
Peter Vrtačnik, Špela Kos, Stephen A Bustin, Janja Marc, Barbara Ostanek
RESUMEN
The accuracy of techniques such as microarrays, reverse transcription polymerase chain reaction, and whole transcriptome shotgun sequencing is critically dependent on RNA quality. We have repeatedly observed extensive RNA degradation following trypsinization, a routine procedure used to dissociate adherent tissue culture cells prior to RNA extraction. This study investigated the cause of this degradation and identifies an alternative procedure that enables extraction of intact high-quality RNA. Trypsinization and several alternative procedures were used to dissociate a range of different cell lines prior to RNA extraction. The contribution of exogenous ribonucleases or induction of endogenous ribonucleases by trypsin reagent proteases to RNA degradation was examined. Trypsinization resulted in a complete degradation of RNA regardless of cell line type, differentiation stage, or passage number. This occurred when intact RNA was incubated directly with trypsin and was not suppressed by inhibiting trypsin's protease activity. Prevention of degradation by sodium hypochlorite treatment of trypsin reagent identified the presence of ribonucleases in trypsin derived from animal pancreas. Consistent extraction of high-quality RNA requires the use of direct cell lysis with a phenol guanidine-based reagent or an animal origin-free protease-based dissociation agent if enzymatic detachment prior to RNA extraction cannot be avoided.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Ethanol, for residue analysis
Supelco
Ethanol, standard for GC
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone and 0.5% 4-methyl-2-pentanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
2-Propanol, analytical standard