• Inicio
  • Resultados de la búsqueda
  • Antioxidant capacity of hydrolyzed protein fractions obtained from an under-explored seed protein: Gingerbread plum (Neocarya macrophylla).
Antioxidant capacity of hydrolyzed protein fractions obtained from an under-explored seed protein: Gingerbread plum (Neocarya macrophylla).
Journal of food science and technology (2015-04-22)
Tidjani Amza, Issoufou Amadou, Abdourahamane Balla, HuiMing Zhou
RESUMEN
The antioxidant capacity of gingerbread plum kernel protein fraction (albumin, globulin and glutelin) hydrolysates (GPKH) was studied. Gingerbread plum kernel protein fractions were hydrolyzed through a combined action of two digestive enzymes (pespsin and trypsin). The hydrolyzed fractions were subjected to antioxidant test via several chemical assays such as: DPPH (2,2-diphenyl-1-picrylhydrazyl) radical scavenging activity, hydroxyl radical-scavenging activity, reducing power and metal chelating activity. Total phenolic contents, amino acid composition and molecular weight distribution were also evaluated. The glutelin fraction hydrolysate showed the strongest antioxidative activity throughout the entire investigation: 79.09, 58.81, 52.08 % and 40.7 μg/mL GAE for DPPH, hydroxyl radical, chelating activity and total phenolics respectively. GPKH possess a molecular weight ranging from 300 to 4000 Da and also showed much more high reducing power than some common standards such as BHA and α-tocopherol indicating that, hydrolysates derived from gingerbread plum kernel protein could be a new antioxidants source.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder (may contain lumps)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
Gallic acid, certified reference material, TraceCERT®
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Phthaldialdehyde, for fluorescence, ≥99.0% (HPLC)
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Octylphosphonic acid, 97%