  • Search Results
  • Consequences of the matrix effect on recovery of dinotefuran and its metabolites in green tea during tandem mass spectrometry analysis.
Consequences of the matrix effect on recovery of dinotefuran and its metabolites in green tea during tandem mass spectrometry analysis.
Food chemistry (2014-08-31)
Md Musfiqur Rahman, A M Abd El-Aty, Jeong-Heui Choi, Sung-Woo Kim, Sung Chul Shin, Jae-Han Shim
ABSTRACT
Determining the residues of dinotefuran and its metabolites (MNG, UF, and DN) is highly problematic because of their polar characteristics. Additionally, tea contains many compounds that can interfere with residue analysis. Thus, the aim of the present study was to refine the extraction method that assures good recoveries for dinotefuran and its metabolites and removes most of the matrix components in green tea using liquid chromatography-tandem mass spectrometry (LC/MS/MS). We attempted to increase the extraction efficiency of the QuEChERS method by selecting the appropriate solvents among ethyl acetate, acetone, isopropanol, 25% methanol in acetonitrile, and methanol. We found that methanol was the best extraction solvent for dinotefuran and its polar metabolites in dry green tea samples; however, due to a limitation of an appropriate partitioning salt, acetonitrile was used as the extraction solvent. Matrix enhancement and suppression effects were observed for all analytes, which made the recovery rates variable. DN recovery was <70% when compared with matrix-matched calibration, whereas it was within the acceptable range (70-120%) when compared with solvent calibration. The opposite was observed for MNG and dinotefuran due to a matrix suppression effect. UF recovery was consistent in both matrix-matched and solvent calibrations despite having little suppressive effect. The method was successfully applied and dinotefuran and its metabolite residues were found in field-incurred green tea samples. The current findings suggest that using methanol as an appropriate QuEChERS solvent for problematic polar pesticides and investigating a suitable partitioning salt would considerably strengthen the practical impact of such data.
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, suitable for HPLC, contains 10 % (v/v) water, 0.1 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, analytical standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)