Multi-variable operational characteristic studies of on-column oxidative protein refolding at high loading concentrations.
Journal of chromatography. A (2014-08-06)
Pegah Saremirad, Jeffery A Wood, Yan Zhang, Ajay K Ray
RESUMEN
Chromatographic-based protein refolding techniques have proven to be superior to conventional dilution refolding methods, due to the higher loading concentration and simultaneous purification. Among these techniques, Size Exclusion Chromatography (SEC) has in particular been demonstrated as an effective method for refolding of variety of proteins. To date existing studies of protein refolding at high concentrations (>1mg/mL) in SEC have primarily been conducted as single factor studies, in which a single parameter is varied to assess impact on operating performance, which does not allow for determination of the interactions of different operating parameters and optimized operating conditions. In this work a multi-variable investigation of size exclusion protein refolding at high protein concentration using lysozyme as a model protein was performed, in order to quantify the interaction of factors and optimize performance. It was observed when l-arginine is used as an additive the refolding yield becomes independent of the protein concentration and refolding buffer pH, providing that a redox couple is used to assist the reformation of disulfide bridges. Furthermore, the pore accessibility for small molecules was reduced at the presence of this additive particularly at higher protein concentrations indicating slower removal of these molecules and a possible additional mechanism of aggregation prevention. Using the subsequent optimized refolding buffer, a refolding yield of more than 90% was obtained for up to 40mg/mL loading concentration of lysozyme which has only been reported for a urea gradient SEC (8-2M) with lower equilibration and elution flow rates due to high viscosity of buffer containing high concentrations of urea.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
SAFC
L-Cysteine
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture