Rapid electrical stimulation causes alterations in cardiac intercellular junction proteins of cardiomyocytes.
American journal of physiology. Heart and circulatory physiology (2014-03-13)
Tadamitsu Nakashima, Tomoko Ohkusa, Yoko Okamoto, Masaaki Yoshida, Jong-Kook Lee, Yoichi Mizukami, Masafumi Yano
RESUMEN
The intercellular junctions contain two complexes, adhesion junctions (AJ) and connexin (Cx) gap junctions (GJs). GJs provide the pathway for intercellular current flow. AJs mediate normal mechanical coupling and play an important role in the stability of GJs. We investigated the effects of rapid electrical stimulation (RES) on cardiac intercellular junctions, especially β-catenin and Cx43 alterations. We also studied the effects of ANG II receptor blockade on intercellular junction remodeling. Neonatal rats were euthanized by decapitation, and cardiomyocytes were prepared, cultured, and subjected to RES. We used real-time PCR, western blot analysis, and immunohistochemical methods. Conduction properties were examined by an extracellular potential mapping system. Cx43 protein expression in cardiomyocytes was significantly increased after 60 min. β-Catenin expression in the total cell fraction was significantly increased after 30 min. The expression level of β-catenin in the nucleus, which functions as a T cell factor/lymphocyte enhancer binding factor transcriptional activator of Cx43 with its degradation regulated by glycogen synthase kinase-3β, was dramatically increased after 10 min. Conduction velocity was increased significantly by RES for 60 min. Olmesartan prevented most these effects of RES. We showed an increase of phosphorylated glycogen synthase kinase-3β, which is phosphorylated by activated MAPKs and inhibits β-catenin degradation, was attenuated by olmesartan. The changes in β-catenin precede Cx43 GJ remodeling and might play an important role in the formation and stability of GJs. Olmesartan might be a new upstream arrhythmia therapy by modulating intercellular junction remodeling through the β-catenin signaling pathway.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−FITC antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)–Peroxidase antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG, Native−Peroxidase antibody, Mouse monoclonal antibody produced in mouse, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG, Native antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)−FITC antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-GAPDH antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-GAPDH antibody produced in mouse, clone 3C2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (Fc specific)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (γ-chain specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in sheep, affinity isolated antibody, buffered aqueous solution