• Inicio
  • Resultados de la búsqueda
  • Neonatal glucocorticoid treatment: long-term effects on the hypothalamus-pituitary-adrenal axis, immune system, and problem behavior in 14-17 year old adolescents.
Neonatal glucocorticoid treatment: long-term effects on the hypothalamus-pituitary-adrenal axis, immune system, and problem behavior in 14-17 year old adolescents.
Brain, behavior, and immunity (2014-12-03)
Maike Ter Wolbeek, Annemieke Kavelaars, Willem B de Vries, Marijke Tersteeg-Kamperman, Sylvia Veen, René F Kornelisse, Mirjam van Weissenbruch, Wim Baerts, Kian D Liem, Frank van Bel, Cobi J Heijnen
RESUMEN
Neonatal glucocorticoid (GC) treatment is used to prevent bronchopulmonary dysplasia (BPD) in prematurely born babies. In the 1990s, treatment regimens with relatively high doses of dexamethasone (DEX) were common. As an alternative, hydrocortisone (HC) was used. Earlier, we compared long-term effects of both GCs in children aged 7-10 and detected adverse effects of neonatal DEX treatment, but not of HC, on a range of outcomes. The aim of the current cohort study was to investigate whether long-term effects of neonatal DEX were maintained and whether effects of HC remained absent at adolescent age (14-17years). We compared 71 DEX-treated and 67 HC-treated adolescents. In addition, 71 adolescents who were not neonatally treated with GCs participated. All were born <32weeks of gestation. DEX-treated girls showed increased adrenocorticotropic hormone (ACTH) and cortisol responses in the Trier Social Stress Test. The cortisol awakening response was lower in HC-treated participants compared to untreated participants. Negative feedback function of the HPA-axis in the dexamethasone suppression test did not differ between groups. In contrast to our observations at the age of 7-10years, we did not observe group differences in mitogen-induced cytokine production at the age of 14-17years. DEX-treated girls showed more social problems and anxious/depressed behavior than HC-treated girls. Untreated girls showed more problem behavior as well. In conclusion, our results suggest that, especially in girls, neonatal DEX has a programming effect on the HPA-axis and on the ability to adjust to the environment. The loss of group differences on immune system measures indicate that potentially negative effects detected at a younger age subsided.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard