CHRNB3 c.-57A>G functional promoter change affects Parkinson's disease and smoking.
Neurobiology of aging (2014-04-16)
Anat Bar-Shira, Mali Gana-Weisz, Ziv Gan-Or, Eytan Giladi, Nir Giladi, Avi Orr-Urtreger
RESUMEN
Cigarette smoking is protective in Parkinson's disease (PD), possibly because of nicotine action on brain nicotinic-acetylcholine receptors. The β3 nicotinic-acetylcholine receptor subunit (encoded by CHRNB3) is depleted in the striatum of PD patients and associated with nicotine dependence. Herein, the CHRNB3 gene was sequenced, and the c.-57G allele frequency was 0.31 and 0.26 among patients (n = 596) and controls (n = 369), respectively (p = 0.02, odds ratio = 1.33, 95% confidence interval = 1.03-1.73). The c.-57G allele was strongly associated with smoking in patients, as 48.4% of c.-57G carriers compared with 32.6% of noncarriers reported smoking history (p < 0.0001). The transcription factor Oct-1 binding was almost eliminated in lymphoblasts with the c.-57G/G genotype, to only 6.5% percent, and the CHRNB3 promoter activity was reduced in cells with the c.-57G/G genotype by 96%-70%. These findings suggest that the CHRNB3 c.-57A>G alteration affects the promoter activity and is associated with PD and smoking in PD patients. It is therefore possible that nicotine may be valuable for patients who carry this alteration and beneficial in PD only for patients with specific genotypes.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone GG-7, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone HP-6017, ascites fluid
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal, clone HP-6017, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Goat/Sheep IgG−Peroxidase antibody, Mouse monoclonal antibody produced in mouse, clone GT-34, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)–Peroxidase antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
Sigma-Aldrich
Anti-Human IgG (Fc specific)−FITC antibody, Mouse monoclonal antibody produced in mouse, clone HP-6017, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Goat/Sheep IgG antibody produced in mouse, clone GT-34, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG, Native−Peroxidase antibody, Mouse monoclonal antibody produced in mouse, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Human IgG (Fab specific) antibody produced in mouse, clone GG-6, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG, Native antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Human IgG2 (Fc specific) antibody produced in mouse, clone HP-6002, ascites fluid
Sigma-Aldrich
Anti-Goat/Sheep IgG−Biotin antibody, Mouse monoclonal, clone GT-34, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Goat/Sheep IgG−Alkaline Phosphatase antibody, Mouse monoclonal, clone GT-34, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Goat/Sheep IgG−FITC antibody, Mouse monoclonal antibody produced in mouse, clone GT-34, purified from hybridoma cell culture
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Monoclonal Anti-Human IgG4 (pFc′ specific) antibody produced in mouse, clone HP-6023, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Human IgG (γ-chain specific) antibody produced in mouse, clone GG-5, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Rat IgG2a antibody produced in mouse, clone R2A-2, ascites fluid