Merck
Core-shell silver nanoparticles for optical labeling of cells.
Analytical biochemistry (2014-04-24)
Kyle D Dukes, Kenneth A Christensen, George Chumanov
ABSTRACT
Silver nanoparticles have been modified with self-assembled monolayers of hydroxyl-terminated long chain thiols and encapsulated with a silica shell. The resulting core-shell nanoparticles were used as optical labels for cell analysis using flow cytometry and microscopy. The excitation of plasmon resonances in nanoparticles results in strong depolarized scattering of visible light, permitting detection at the single nanoparticle level. The nanoparticles were modified with neutravidin via epoxide-azide coupling chemistry, to which biotinylated antibodies targeting cell surface receptors were bound. The nanoparticle labels exhibited long-term stability in solutions with high salt concentrations without aggregation or silver etching. Labeled cells exhibited two orders of magnitude enhancement of the scattering intensity compared with unlabeled cells.
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.9%
Sigma-Aldrich
1-Undecanol, 99%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
(3-Glycidyloxypropyl)trimethoxysilane, ≥98%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Undecyl alcohol, 97%, FG
Sigma-Aldrich
Tetramethyl orthosilicate, ≥99%
Sigma-Aldrich
Tetramethyl orthosilicate, 98%
Sigma-Aldrich
Tetramethyl orthosilicate, purum, ≥98.0% (GC)
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetramethyl orthosilicate, deposition grade, ≥98%, ≥99.9% trace metals basis
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%