  • Search Results
  • Cellular uptake kinetics of bortezomib in relation to efficacy in myeloma cells and the influence of drug transporters.
Cellular uptake kinetics of bortezomib in relation to efficacy in myeloma cells and the influence of drug transporters.
Cancer chemotherapy and pharmacology (2014-12-06)
Jannick Clemens, Anja Seckinger, Dirk Hose, Dirk Theile, Magdalena Longo, Walter Emil Haefeli, Jürgen Burhenne, Johanna Weiss
ABSTRACT
Despite overall successful application to multiple myeloma patients, clinical efficacy of the proteasome inhibitor bortezomib is typically challenged by primary and secondary resistance of unknown origin. So far, the potential impact of intracellular concentrations on drug efficacy of bortezomib and the influence of drug transporters are unknown. We determined cellular bortezomib kinetics in nine myeloma cell lines using ultrahigh-performance liquid chromatography coupled to tandem mass spectrometry. The potential influence of drug transporters on the uptake kinetics observed in these cell lines was investigated by testing substrate characteristics of bortezomib for several transporters in over-expressing model cells. Additionally, transporter mRNA expression was quantified in myeloma cell lines by real-time polymerase chain reaction (RT-PCR). All myeloma cells revealed an extensive intracellular bortezomib accumulation (47.5-183 ng/ml) exceeding extracellular concentrations (0.04-0.17 ng/ml) by more than factor 1,000. Only organic anion-transporting polypeptide 1B1 facilitated the uptake in over-expressing cells, however, to a negligible extent (factor 1.36). Bortezomib efflux via P-glycoprotein was confirmed by demonstrating reduced sensitivity (IC50 11.6 vs. 2.8 ng/ml) and intracellular concentrations (-56.1%) in over-expressing cells compared to controls. RT-PCR revealed a varying but overall weak transporter expression in the studied myeloma cells without any correlation to intracellular concentrations. Although principally valid as demonstrated in the P-glycoprotein over-expressing cell model, there was no significant correlation between intracellular concentrations and bortezomib efficacy in myeloma cell lines. Differences in intracellular concentrations in myeloma cell lines neither result from variable transporter expression nor represent the main factor determining bortezomib efficacy in vitro.
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Rifampicin, ≥97% (HPLC), powder
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Glycerol, FCC, FG
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone and 0.5% 4-methyl-2-pentanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Hydrogen chloride, puriss., ≥99.8%