RAD001 enhances the radiosensitivity of SCC4 oral cancer cells by inducing cell cycle arrest at the G2/M checkpoint.
Anticancer research (2014-06-13)
Chih-Chia Yu, Shih-Kai Hung, Hui-Fen Liao, Ching-Chih Lee, Hon-Yi Lin, Hung-Chih Lai, Szu-Chin Li, Hsu-Chueh Ho, Hsien-Bin Huang, Yu-Chieh Su
RESUMEN
Inhibition of mammalian target of rapamycin (mTOR) kinase enhances the radiosensitivity of some cancer cells. We investigated the effect of RAD001, an mTOR inhibitor, on irradiated oral cancer cell lines. Clonogenic assays were performed to determine the radiosensitivity of SCC4 and SCC25 cells after treatment with RAD001. Target protein phosphorylation, apoptosis, and cell-cycle progression were assessed in SCC4 cells treated with RAD001 with and without ionizing radiation. RAD001 increased the radiosensitivity of SCC4 cells without affecting cell death; it also inhibited phosphorylation of mTOR, S6, and factor 4E binding protein 1 and reduced the clonogenic survival of irradiated cancer cells. RAD001 combined with radiation increased G2 arrest by activating CHK1, which phosphorylates CDC25C at Ser216, thereby inhibiting CDC2-cyclin B 1 complex formation. RAD001 enhances the radiosensitivity of SCC4 cells by inhibiting mTOR signaling and inducing G2 cell-cycle arrest through disruption of the G2 checkpoint.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)–Peroxidase antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG, Native−Peroxidase antibody, Mouse monoclonal antibody produced in mouse, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG, Native antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture