• Inicio
  • Resultados de la búsqueda
  • Enhanced anaerobic biodegradation of OCDD-contaminated soils by Pseudomonas mendocina NSYSU: microcosm, pilot-scale, and gene studies.
Enhanced anaerobic biodegradation of OCDD-contaminated soils by Pseudomonas mendocina NSYSU: microcosm, pilot-scale, and gene studies.
Journal of hazardous materials (2014-07-06)
Y T Tu, J K Liu, W C Lin, J L Lin, C M Kao
RESUMEN
In this study, microcosm and pilot-scale experiments were performed to investigate the capability and effectiveness of Pseudomonas mendocina NSYSU (P. mendocina NSYSU) on the bioremediation of octachlorodibenzo-p-dioxin (OCDD)-contaminated soils. The objectives were to evaluate the (1) characteristics of P. mendocina NSYSU, (2) feasibility of enhancing OCDD biodegradation with the addition of P. mendocina NSYSU and lecithin, and (3) variation in microbial diversity and genes responsible for the dechlorination of OCDD. P. mendocina NSYSU was inhibited when salinity was higher than 7%, and it could biodegrade OCDD under reductive dechlorinating conditions. Lecithin could serve as the solubilization agent causing the enhanced solubilization and dechlorination of OCDD. Up to 71 and 62% of OCDD could be degraded after 65 days of incubation under anaerobic conditions with and without the addition of lecithin, respectively. Decreased OCDD concentrations caused significant increase in microbial diversity. Results from the pilot-scale study show that up to 75% of OCDD could be degraded after a 2.5-month operational period with lecithin addition. Results from the gene analyses show that two genes encoding the extradiol/intradiol ring-cleavage dioxygenase and five genes encoding the hydrolase in P. mendocina NSYSU were identified and played important roles in OCDD degradation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Supelco
Acetone, analytical standard
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sulfuric acid, puriss. p.a., 95-97% (T)