  • Search Results
  • Oral Candida carriage and immune status in Thai human immunodeficiency virus-infected individuals.
Oral Candida carriage and immune status in Thai human immunodeficiency virus-infected individuals.
Journal of medical microbiology (2014-03-05)
Panida Thanyasrisung, Piyanate Kesakomol, Patchara Pipattanagovit, Pornpan Youngnak-Piboonratanakit, Waranuch Pitiphat, Oranart Matangkasombut
ABSTRACT
Oral candidiasis is a common opportunistic infection among human immunodeficiency virus (HIV)-infected individuals, with growing concerns about the emergence of non-albicans species with resistance to antifungal agents. This cross-sectional study determined the prevalence of oral Candida species in Thai HIV-infected adults and factors affecting their colonization. Candida species were identified from oral rinse samples of 60 HIV-infected participants of the MTCT-Plus initiative and 49 healthy controls by culture-based and molecular assays. The prevalence of oral Candida carriage was similar in HIV-infected patients (56.6 %) and in controls (55.1 %, P = 0.87). Candida albicans was the most predominant species in both groups (94.1 % of Candida carriers in HIV, 88.9 % in control). Interestingly, Candida dubliniensis was the second most common species in controls (29.6 %) and the third in HIV-infected patients (11.8 %, P = 0.08). Multivariate analysis showed that, amongst HIV-infected individuals, CD4 count <200 cells mm(-3) was associated with increased prevalence of oral carriage of both C. albicans (P = 0.03) and non-albicans species (P = 0.03). Moreover, patients with tuberculosis infection had a higher prevalence of the non-albicans species than those without (P = 0.03). Intriguingly, contraceptive use was also associated positively with non-albicans and multi-species carriage (P = 0.04 for both). However, use of antiretroviral drugs protected the patients from Candida carriage (P = 0.03), especially from C. albicans (P = 0.02). In conclusion, while HIV-infected individuals had a similar prevalence of oral Candida carriage to that of the control group, host immune status, tuberculosis infection, and contraceptive use may influence oral colonization of Candida, especially of the non-albicans species.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard