  • Search Results
  • Membrane-anchored human Rab GTPases directly mediate membrane tethering in vitro.
Membrane-anchored human Rab GTPases directly mediate membrane tethering in vitro.
Biology open (2014-11-02)
Naoki Tamura, Joji Mima
ABSTRACT
Rab GTPases are master regulators of eukaryotic endomembrane systems, particularly functioning in membrane tethering to confer the directionality of intracellular membrane trafficking. However, how exactly Rab GTPases themselves act upon membrane tethering processes has remained enigmatic. Here, we thoroughly tested seven purified Rab GTPases in human, which localize at the various representative organelles, for their capacity to support membrane tethering in vitro. Strikingly, we found that three specific human Rabs (endoplasmic reticulum/Golgi Rab2a, early endosomal Rab5a, and late endosomal/lysosomal Rab7a) strongly accelerated membrane aggregation of synthetic liposomes even in the absence of any additional components, such as classical tethers, tethering factors, and Rab effectors. This Rab-induced membrane aggregation was a reversible membrane tethering reaction that can be strictly controlled by the membrane recruitment of Rab proteins on both apposing membranes. Thus, our current reconstitution studies establish that membrane-anchored human Rab GTPases are an essential tethering factor to directly mediate membrane tethering events.
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M