  • Search Results
  • Simultaneous determination of neonicotinoid insecticides in human serum and urine using diatomaceous earth-assisted extraction and liquid chromatography-tandem mass spectrometry.
Simultaneous determination of neonicotinoid insecticides in human serum and urine using diatomaceous earth-assisted extraction and liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-08-30)
Tadashi Yamamuro, Hikoto Ohta, Mika Aoyama, Daisuke Watanabe
ABSTRACT
A rapid and sensitive analytical method was developed for simultaneous determination of eight neonicotinoid insecticides (acetamiprid, clothianidin, dinotefuran, flonicamid, imidacloprid, nitenpyram, thiacloprid and thiamethoxam) and three specific metabolites of acetamiprid (N-desmethylacetamiprid, 5-(N-acetyl-N-methylaminomethyl)-2-chloropyridine and 5-(N-acetylaminomethyl)-2-chloropyridine) in human serum and urine. A diatomaceous earth-assisted extraction using Extrelut NT3 column with chloroform/2-propanol (3:1, v/v) as eluent was selected for the single step cleanup procedure for all the target compounds. Qualitative and quantitative analyses were conducted by liquid chromatography-tandem mass spectrometry with multiple reaction monitoring mode. The limits of detection and the limits of quantification of eleven compounds were in the ranges of 0.1-0.2ng/mL and 0.5-10ng/mL for serum, 0.1-1ng/mL and 1-10ng/mL for urine, respectively. The extraction recoveries were between 80.9% and 101.8% for serum samples, 91.9% and 106% for urine samples. The intra-day RSDs and the inter-day RSDs were less than 11.5% and 13.2% for serum, less than 8.3% and 8.8% for urine. The proposed procedure will be suitable for forensic investigations of human poisoning cases with neonicotinoid insecticides. This is the first report of simultaneous determination of eight neonicotinoids in serum and urine samples.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Supelco
Thiamethoxam-d3, PESTANAL®, analytical standard
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Thiacloprid, PESTANAL®, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Dinotefuran, PESTANAL®, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%