  • Search Results
  • Serial coupling of ion-exchange and size-exclusion chromatography to determine aggregation levels in mAbs in the presence of a proteinaceous excipient, recombinant human serum albumin.
Serial coupling of ion-exchange and size-exclusion chromatography to determine aggregation levels in mAbs in the presence of a proteinaceous excipient, recombinant human serum albumin.
Journal of pharmaceutical sciences (2014-12-03)
Paul Luigi Gargani Weisbjerg, Mikael Bjerg Caspersen, Ken Cook, Marco Van De Weert
ABSTRACT
Aggregation levels of therapeutic proteins may be difficult to determine in mixtures containing other proteinaceous excipients. We performed a feasibility study of using serial coupling of an anion exchange and size exclusion column to determine the aggregation levels of four different model monoclonal antibodies (mAb) mixed with the model proteinaceous excipient recombinant human serum albumin (rHSA). For three of the four mAbs suitable elution conditions could be established. From the limitations imposed by the pI of the rHSA, the pI of the mAb and the nature of the columns used, it was possible to propose a set of general conditions that allows quantification of the aggregation level of a therapeutic protein in the presence of a proteinaceous excipient: The excipient protein and protein of interest should differ in pI by a minimum of 0.5 units, and the pI of the protein of interest should not be higher than ca. 8.5.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material