  • Search Results
  • MTHFR-Ala222Val and male infertility: a study in Iranian men, an updated meta-analysis and an in silico-analysis.
MTHFR-Ala222Val and male infertility: a study in Iranian men, an updated meta-analysis and an in silico-analysis.
Reproductive biomedicine online (2015-09-19)
Hossein Nikzad, Mohammad Karimian, Kobra Sareban, Maryam Khoshsokhan, Abasalt Hosseinzadeh Colagar
ABSTRACT
Methylenetetrahydrofolate reductase (MTHFR) functions as a main regulatory enzyme in folate metabolism. The association of MTHFR gene Ala222Val polymorphism with male infertility in an Iranian population was investigated by undertaking a meta-analysis and in-silico approach. A genetic association study included 497 men; 242 had unexplained infertility and 255 were healthy controls. Polymerase chain reaction restriction fragment length polymorphism was used for genotyping MTHFR-Ala222Val. OpenMeta[Analyst] software was used to conduct the analysis; 22 studies were identified by searching PubMed and the currently reported genetic association study. A novel in-silico approach was used to analyse the effects of Ala222Val substitution on the structure of mRNA and protein. Genetic association study revealed a significant association of MTHFR-222Val/Val genotype with oligozoospermia (OR 2.32; 95% CI, 1.12 to 4.78; P = 0.0451) and azoospermia (OR 2.59; 95% CI 1.09 to 6.17; P = 0.0314). Meta-analysis for allelic, dominant and codominant models showed a significant association between Ala222Val polymorphism and the risk of male infertility (P < 0.001). In silico-analysis showed MTHFR-Ala222Val affects enzyme structure and could also change the mRNA properties (P = 0.1641; P < 0.2 is significant). The meta-analysis suggested significant association of MTHFR-Ala222Val with risk of male infertility, especially in Asian populations.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M