  • Search Results
  • Human health risk assessment of heavy metals in tropical fish and shellfish collected from the river Buriganga, Bangladesh.
Human health risk assessment of heavy metals in tropical fish and shellfish collected from the river Buriganga, Bangladesh.
Environmental science and pollution research international (2015-06-06)
Md Kawser Ahmed, Mohammad Abdul Baki, Md Saiful Islam, Goutam Kumar Kundu, Md Habibullah-Al-Mamun, Santosh Kumar Sarkar, Md Muzammel Hossain
ABSTRACT
Although fish, crustacean, and shellfish are significant sources of protein, they are currently affected by rapid industrialization, resulting in increased concentrations of heavy metals. Accumulation of heavy metals (V, Cr, Mn, Ni, Cu, Zn, As, Se, Mo, Ag, Cd, Sb, Ba, and Pb) and associated human health risk were investigated in three fish species, namely Ailia coila, Gagata youssoufi, and Mastacembelus pancalus; one crustacean (prawn), Macrobrachium rosenbergii; and one Gastropoda, Indoplanorbis exustus, collected from the Buriganga River, Bangladesh. Samples were collected from the professional fishermen. Cu was the most accumulated metal in M. rosenbergii. Ni, As, Ag, and Sb were in relatively lower concentrations, whereas relatively higher accumulation of Cr, Mn, Zn, and Se were recorded. Mn, Zn, and Pb were present in higher concentrations than the guidelines of various authorities. There were significant differences in metal accumulation among different fish, prawn, or shellfish species. Target hazard quotient (THQ) and target cancer risk (TR) were calculated to estimate the non-carcinogenic and carcinogenic health risks, respectively. The THQ for individual heavy metals were below 1 suggesting no potential health risk. But combined impact, estimated by hazard index (HI), suggested health risk for M. pancalus consumption. Although consumption of fish at current accumulation level is safe but continuous and excess consumption for a life time of more than 70 years has probability of target cancer risk.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%