  • Search Results
  • Korean Red Ginseng Extract Enhances the Anticancer Effects of Imatinib Mesylate Through Abrogation p38 and STAT5 Activation in KBM-5 Cells.
Korean Red Ginseng Extract Enhances the Anticancer Effects of Imatinib Mesylate Through Abrogation p38 and STAT5 Activation in KBM-5 Cells.
Phytotherapy research : PTR (2015-04-11)
Sang Yoon Jung, Chulwon Kim, Wan-Seok Kim, Seok-Geun Lee, Jun-Hee Lee, Bum Sang Shim, Sung-Hoon Kim, Kyoo Seok Ahn, Kwang Seok Ahn
ABSTRACT
Although imatinib mesylate (IM) in the treatment of chronic myelogenous leukemia (CML) remains the best example of successful targeted therapy, majority of patients with CML suffer its toxicity profile and develop chemoresistance to existing therapeutic agents. Thus, there is a need to develop novel alternative therapies for the treatment of CML. Here, we investigated whether Korean red ginseng extract (KRGE) could suppress the proliferation and induce chemosensitization in human CML cells. Also, we used a human phospho-antibody array containing 46 antibodies against signaling molecules to examine a subset of phosphorylation events after treatment. Korean red ginseng extract broadly suppressed the proliferation of five different cell lines, but KRGE was found to be the most potent inducer of apoptosis against KBM-5 cells. It also abrogated the expression of Bcl-2 (B-cell lymphoma 2), Bcl-xL (B-cell lymphoma-extra large), survivin, inhibitors of apoptosis protein 1/2, COX-2 (Cyclooxygenase-2), cyclin D1, matrix metalloproteinase-9, and VEGF (Vascular endothelial growth factor), as well as upregulated the expression of pro-apoptotic gene products. Interestingly, KRGE also enhanced the cytotoxic and apoptotic effect of IM in KBM-5 cells. The combination treatment of KRGE and IM caused pronounced suppression of p38 and signal transducer and activator of transcription 5 phosphorylation and induced phosphorylation of p53 compared with the individual treatment. Our results demonstrate that KRGE can enhance the anticancer activity of IM and may have a substantial potential in the treatment of CML.
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate