Macro-hydrogels versus nanoparticles by the controlled assembly of polysaccharides.
Carbohydrate polymers (2015-10-03)
M Costalat, P Alcouffe, L David, T Delair
RESUMEN
The controlled assembly of oppositely charged chitosan (CS, Mw ∼ 33 × 10(3) to 600 × 10(3)g mol(-1)) and dextran sulfate (DS, Mw = 1.3 × 10(6)g mol(-1)) or heparin (HP, Mw = 1.8 × 10(4)g mol(-1)) led either to nanoparticles or macro-hydrogels, at room temperature. The control over the electrostatic attractive interactions was achieved using 2 mol L(-1) NaCl in the polyion solutions and subsequent dialysis to let the assembly occur. Macrohydrogels formed with an excess of polyanion. In the presence of an excess of polycation, colloidal gels were exclusively obtained. At salt concentrations lower than 1 mol L(-1), the spontaneous gelation provided macro-hydrogels, whatever the polyion in excess. Rheology measurements showed a similar elastic behaviour for CS-DS and CS-HP hydrogels, though CS-HP hydrogels appeared less cohesive. SAXS experiments revealed an aggregate morphology with internal and surface structure depending on the degree of acetylation (DA) of chitosan.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture