  • Search Results
  • GlxA is a new structural member of the radical copper oxidase family and is required for glycan deposition at hyphal tips and morphogenesis of Streptomyces lividans.
GlxA is a new structural member of the radical copper oxidase family and is required for glycan deposition at hyphal tips and morphogenesis of Streptomyces lividans.
The Biochemical journal (2015-07-25)
Amanda K Chaplin, Marloes L C Petrus, Giulia Mangiameli, Michael A Hough, Dimitri A Svistunenko, Peter Nicholls, Dennis Claessen, Erik Vijgenboom, Jonathan A R Worrall
ABSTRACT
Streptomyces lividans displays a distinct dependence on copper to fully initiate morphological development. Evidence has accumulated to implicate the participation of an extracytoplasmic cuproenzyme in morphogenesis. In the present study, we show that GlxA fulfils all criteria to be that cuproenzyme. GlxA is membrane associated and has an active site consisting of a mononuclear copper and a cross-linked Y-C cofactor. The domain organization of the tertiary structure defines GlxA as a new structural member of the mono-copper oxidase family, with copper co-ordination geometry similar to, but spectroscopically distinct from fungal galactose oxidase (Gox). EPR spectroscopy reveals that the oxidation of cupric GlxA generates a protein radical residing on the Y-C cross-link. A variety of canonical Gox substrates (including D-galactose) were tested but none were readily turned over by GlxA. A glxA null-mutant leads to loss of glycan accumulation at hyphal tips and consequently a drastically changed morphology both on solid substrates and in liquid-grown environments, a scenario similarly observed in the absence of the neighbouring glycan synthase CslA (cellulase synthase-like protein). In addition the glxA mutant has lost the stimulation of development by copper, supporting a model whereby the enzymatic action of GlxA on the glycan is required for development and morphology. From a biotechnology perspective, the open mycelium morphology observed with the glxA mutant in submerged culture has implications for use as an enzyme production host.
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Sodium hydrosulfite, ≥82% (RT)
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
Glyoxal solution, BioReagent, for molecular biology, ~40% in H2O (~8.8 M)
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Glyoxal solution, 40 wt. % in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
D-(+)-Galactose, BioXtra, pH 5.0-7.0 (20 °C, 1 M in H2O), ≥99%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Galactose, ≥99%
Sigma-Aldrich
D-(+)-Galactose, powder, anhydrous, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥98%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)