• Inicio
  • Resultados de la búsqueda
  • Quantitation of various indolinyl caged glutamates as their o-phthalaldehyde derivatives by high performance liquid chromatography coupled with tandem spectroscopic detections: derivatization, stoichiometry and stability studies.
Quantitation of various indolinyl caged glutamates as their o-phthalaldehyde derivatives by high performance liquid chromatography coupled with tandem spectroscopic detections: derivatization, stoichiometry and stability studies.
Journal of chromatography. A (2015-04-05)
A Vasanits-Zsigrai, O Majercsik, G Tóth, A Csámpai, Cs Haveland-Lukács, D Pálfi, Z Szadai, B Rózsa, I Molnár-Perl
RESUMEN
Quantification, stability and unique spectroscopic properties of indolinyl-caged glutamates (ICGs), with the o-phthalaldehyde-3-mercaptopropionic acid (OPA-MPA) reagent, were described, at first. As new principle to the field, reactivity and stoichiometry of variously substituted OPA-MPA derivatized ICGs, such as 4-methoxy-7-nitroindolinyl-(MNI-Glu), 4-methoxy-5,7-dinitroindolinyl-(DNI-Glu), 2-dimethylamino-propoxy and dimethylamino-isobutoxy alternatives (2DMA-1PO-DNI-Glu, 1DMA-2P-DNI-Glu and 3DMA-1iBU-DNI-Glu), was demonstrated. Derivatives' stability was determined using high performance liquid chromatography (HPLC) applying simultaneous photodiode array (DAD) and fluorescence (Fl) detections, while their structural identity was confirmed by HPLC-time of flight mass spectrometry (HPLC-TOF-MS). The SH-additive of the reagents was also varied. ICGs react unequivocally, with one OPA-SH-group molecule, in the molar ratios of ([OPA-SH-additive]/[ICG]=1/1, resulting in species with the characteristic isoindole spectral property (EEx/EEm=337/454nm; λmax=337nm). ICGs' isoindole derivatives, due to their sandwich structure, are manifesting the π-π-stacking phenomenon: they fail to show fluorescence. ICGs' stability decreased in the order of MNI-Glu, 2DMA-1PO/1DMA-2PO, 3DMA-1iBU and DNI-Glu, correspondingly, resulting in increasing order of free glutamic acid (GA), as their decomposition product. GA and ICGs were determined as their OPA/MPA derivatives while uncaged species (MNI, DNI and its substituted alternatives) in their initial forms. The practical utility of the method was confirmed analyzing ICGs and their decomposition products, simultaneously. Quantifications' reliability and reproducibility were characterized with the relative standard deviation percentages of responses (RSDs%): for GA 0.41-12 RSD% for ICGs 0.057-7.0 RSD% were obtained. Stability properties of variously substituted, recently introduced ICGs, prepared in laboratories of Institute of Experimental Medicine, were defined.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, suitable for cell culture, BioReagent
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder (may contain lumps)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder
Sigma-Aldrich
Ethanethiol, 97%
Sigma-Aldrich
Phthaldialdehyde, for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Octylphosphonic acid, 97%
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethanethiol, ≥97%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Supelco
Methanol solution, suitable for HPLC, contains 10 % (v/v) water, 0.1 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Ultrapure Acetonitrile