• Inicio
  • Resultados de la búsqueda
  • Elevated Levels of Asymmetric Dimethylarginine (ADMA) in the Pericardial Fluid of Cardiac Patients Correlate with Cardiac Hypertrophy.
Elevated Levels of Asymmetric Dimethylarginine (ADMA) in the Pericardial Fluid of Cardiac Patients Correlate with Cardiac Hypertrophy.
PloS one (2015-08-28)
Zoltan Nemeth, Attila Cziraki, Sandor Szabados, Bernadett Biri, Sandor Keki, Akos Koller
RESUMEN
Pericardial fluid (PF) contains several biologically active substances, which may provide information regarding the cardiac conditions. Nitric oxide (NO) has been implicated in cardiac remodeling. We hypothesized that L-arginine (L-Arg) precursor of NO-synthase (NOS) and asymmetric dimethylarginine (ADMA), an inhibitor of NOS, are present in PF of cardiac patients and their altered levels may contribute to altered cardiac morphology. L-Arg and ADMA concentrations in plasma and PF, and echocardiographic parameters of patients undergoing coronary artery bypass graft (CABG, n = 28) or valve replacement (VR, n = 25) were determined. We have found LV hypertrophy in 35.7% of CABG, and 80% of VR patients. In all groups, plasma and PF L-Arg levels were higher than that of ADMA. Plasma L-Arg level was higher in CABG than VR (75.7 ± 4.6 μmol/L vs. 58.1 ± 4.9 μmol/L, p = 0.011), whereas PF ADMA level was higher in VR than CABG (0.9 ± 0.0 μmol/L vs. 0.7 ± 0.0 μmol/L, p = 0.009). L-Arg/ADMA ratio was lower in the VR than CABG (VRplasma: 76.1 ± 6.6 vs. CABGplasma: 125.4 ± 10.7, p = 0.004; VRPF: 81.7 ± 4.8 vs. CABGPF: 110.4 ± 7.2, p = 0.009). There was a positive correlation between plasma L-Arg and ADMA in CABG (r = 0.539, p = 0.015); and plasma and PF L-Arg in CABG (r = 0.357, p = 0.031); and plasma and PF ADMA in VR (r = 0.529, p = 0.003); and PF L-Arg and ADMA in both CABG and VR (CABG: r = 0.468, p = 0.006; VR: r = 0.371, p = 0.034). The following echocardiographic parameters were higher in VR compared to CABG: interventricular septum (14.7 ± 0.5 mm vs. 11.9 ± 0.4 mm, p = 0.000); posterior wall thickness (12.6 ± 0.3 mm vs. 11.5 ± 0.2 mm, p = 0.000); left ventricular (LV) mass (318.6 ± 23.5 g vs. 234.6 ± 12.3 g, p = 0.007); right ventricular (RV) (33.9 ± 0.9 cm2 vs. 29.7 ± 0.7 cm2, p = 0.004); right atrial (18.6 ± 1.0 cm2 vs. 15.4 ± 0.6 cm2, p = 0.020); left atrial (19.8 ± 1.0 cm2 vs. 16.9 ± 0.6 cm2, p = 0.033) areas. There was a positive correlation between plasma ADMA and RV area (r = 0.453, p = 0.011); PF ADMA and end-diastolic (r = 0.434, p = 0.015) and systolic diameter of LV (r = 0.487, p = 0.007); and negative correlation between PF ADMA and LV ejection fraction (r = -0.445, p = 0.013) in VR. We suggest that elevated levels of ADMA in the PF of patients indicate upregulated RAS and reduced bioavailability of NO, which can contribute to the development of cardiac hypertrophy and remodeling.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99%
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder (may contain lumps)
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder
Sigma-Aldrich
Phthaldialdehyde, for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Octylphosphonic acid, 97%
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99.0% (HPLC)
Sigma-Aldrich
L-Homoarginine hydrochloride, unnatural arginine analog
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, suitable for HPLC, contains 10 % (v/v) water, 0.1 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C