  • Search Results
  • Physicochemical characterization of wet microalgal cells disrupted with instant catapult steam explosion for lipid extraction.
Physicochemical characterization of wet microalgal cells disrupted with instant catapult steam explosion for lipid extraction.
Bioresource technology (2015-05-20)
Jun Cheng, Rui Huang, Tao Li, Junhu Zhou, Kefa Cen
ABSTRACT
Instant catapult steam explosion (ICSE) was employed to disrupt wet microalgal cells for efficient lipid extraction. Physicochemical properties of exploded cells were investigated through SEM, TEM, FTIR, and TGA. The exploded cells increased in fractal dimension (1.53-1.65) when preheat time was prolonged from 0 min to 5 min and in surface pore area when steam pressure was increased. Meanwhile, the exploded cells decreased in mean size (1.69-1.44 μm) when the filling ratio of wet microalgal biomass in the preheat chamber decreased (75-12.5%). Flash evaporation and volume expansion exploded the cell walls and released the cytoplasm of the microalgal cells. These phenomena decreased the carbohydrate content and increased the lipid content in the exploded biomass. However, ICSE treatment did not change the lipid compositions in the microalgal cells. Using isopropanol as a cosolvent significantly increased the yield of lipids extracted with hexane from the exploded wet microalgal biomass.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Methanol solution, suitable for HPLC, contains 10 % (v/v) water, 0.1 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required